CAS RN 51762-67-5
IUPAC Name:
3-nitrobenzene-1,2-dicarbonitrile
Synonyms:
3-nitrobenzene-1,2-dicarbonitrile
Molecular Weight (g/mol):
173.13
Molecular Formula:
C8H3N3O2
InChi Key:
UZJZIZFCQFZDHP-UHFFFAOYSA-N
SMILES:
[O-][N+](=O)C1=CC=CC(C#N)=C1C#N
1
–
1
of
1
results
1
3-Nitrophthalonitrile, 99%
CAS: 51762-67-5 Molecular Formula: C8H3N3O2 Molecular Weight (g/mol): 173.13 MDL Number: MFCD00191558 InChI Key: UZJZIZFCQFZDHP-UHFFFAOYSA-N Synonym: 3-nitrophthalonitrile,2,3-dicyanonitrobenzene,1,2-benzenedicarbonitrile, 3-nitro,3-nitrophthalodinitrile,1,2-dicyano-3-nitrobenzene,3-nitro-1,2-benzenedicarbonitrile,nitrophthalonitrile,3-nitro-phthalonitrile,3-nitrophthalonitrile?,acmc-209kvo PubChem CID: 162652 IUPAC Name: 3-nitrobenzene-1,2-dicarbonitrile SMILES: [O-][N+](=O)C1=CC=CC(C#N)=C1C#N