accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

OHOClCl

CAS RN 51719-65-4

IUPAC Name: 2-(3,5-dichlorophenyl)acetic acid
Synonyms: (3,5-dichlorophenyl)acetic acid
Molecular Weight (g/mol): 205.03
Molecular Formula: C8H6Cl2O2
InChi Key: RERINLRFXYGZEE-UHFFFAOYSA-N
SMILES: OC(=O)CC1=CC(Cl)=CC(Cl)=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results