CAS RN 51693-17-5
CAS RN 51693-17-5
IUPAC Name:
[(2S)-5-oxopyrrolidin-2-yl]methyl 4-methylbenzene-1-sulfonate
Synonyms:
[(2S)-5-oxopyrrolidin-2-yl]methyl 4-methylbenzenesulfonate
Molecular Weight (g/mol):
269.32
Molecular Formula:
C12H15NO4S
InChi Key:
AMZNHHZJURKRFX-JTQLQIEISA-N
SMILES:
CC1=CC=C(C=C1)S(=O)(=O)OC[C@@H]1CCC(=O)N1
1
–
1
of
1
results
1
(S)-(+)-5-(Hydroxymethyl)-2-pyrrolidinone p-toluenesulfonate, 95%
CAS: 51693-17-5 Molecular Formula: C12H15NO4S Molecular Weight (g/mol): 269.32 MDL Number: MFCD00274229,MFCD00274229 InChI Key: AMZNHHZJURKRFX-JTQLQIEISA-N IUPAC Name: [(2S)-5-oxopyrrolidin-2-yl]methyl 4-methylbenzene-1-sulfonate SMILES: CC1=CC=C(C=C1)S(=O)(=O)OC[C@@H]1CCC(=O)N1