CAS RN 5157-89-1
CAS RN 5157-89-1
IUPAC Name:
5-methyl-2-(propan-2-yl)cyclohexyl acetate
Synonyms:
(-)-menthyl acetate dl-MENTHYL ACETATE
Molecular Weight (g/mol):
198.31
Molecular Formula:
C12H22O2
InChi Key:
XHXUANMFYXWVNG-UHFFFAOYNA-N
SMILES:
CC(C)C1CCC(C)CC1OC(C)=O
1
–
1
of
1
results
1
(1S)-(+)-Menthyl acetate, 99%
CAS: 5157-89-1 Molecular Formula: C12H22O2 Molecular Weight (g/mol): 198.306 MDL Number: MFCD00062977 InChI Key: XHXUANMFYXWVNG-DLOVCJGASA-N Synonym: neoisomenthyl acetate,+-menthylacetate,1s-2alpha-1-methylethyl-5alpha-methylcyclohexan-1alpha-ol acetate PubChem CID: 21630934 IUPAC Name: [(1S,2S,5S)-5-methyl-2-propan-2-ylcyclohexyl] acetate SMILES: CC1CCC(C(C1)OC(=O)C)C(C)C