accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 51501-87-2

H3COSiH3CCH3CH3

CAS RN 51501-87-2

IUPAC Name: methoxydimethyl(4-methylphenyl)silane
Synonyms: methoxydimethyl(4-methylphenyl)silane
Molecular Weight (g/mol): 180.32
Molecular Formula: C10H16OSi
InChi Key: GIQXGMLMIZAKLB-UHFFFAOYSA-N
SMILES: CO[Si](C)(C)C1=CC=C(C)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results