accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 51419-59-1

CH3SClOO

CAS RN 51419-59-1

IUPAC Name: (4-methylphenyl)methanesulfonyl chloride
Synonyms: (4-methylphenyl)methanesulfonyl chloride
Molecular Weight (g/mol): 204.67
Molecular Formula: C8H9ClO2S
InChi Key: JALKUHLLMWYIAT-UHFFFAOYSA-N
SMILES: CC1=CC=C(CS(Cl)(=O)=O)C=C1
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)