accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 51417-13-1

CH3NOH3CBr

CAS RN 51417-13-1

IUPAC Name: 5-bromo-1,3-dimethyl-1,2-dihydropyridin-2-one
Synonyms: 5-bromo-1,3-dimethylpyridin-2-one
Molecular Weight (g/mol): 202.05
Molecular Formula: C7H8BrNO
InChi Key: UEHHXNZKBMALIY-UHFFFAOYSA-N
SMILES: CN1C=C(Br)C=C(C)C1=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results