accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

H3CNCH3H3CONN(E)

CAS RN 51325-91-8

IUPAC Name: 2-{2-[(1E)-2-[4-(dimethylamino)phenyl]ethenyl]-6-methyl-4H-pyran-4-ylidene}propanedinitrile
Synonyms: 2-{2-[(1E)-2-[4-(dimethylamino)phenyl]ethenyl]-6-methylpyran-4-ylidene}propanedinitrile
Molecular Weight (g/mol): 303.37
Molecular Formula: C19H17N3O
InChi Key: YLYPIBBGWLKELC-RMKNXTFCSA-N
SMILES: CN(C)C1=CC=C(\C=C\C2=CC(C=C(C)O2)=C(C#N)C#N)C=C1
Grade 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results