accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

ClNNH

CAS RN 51304-58-6

IUPAC Name: 4-phenylpiperidine-4-carbonitrile hydrochloridyl
Synonyms: 4-phenylpiperidine-4-carbonitrile hydrochloridyl
Molecular Weight (g/mol): 221.71
Molecular Formula: C12H14ClN2
InChi Key: RXZQAWBXAWPSJL-UHFFFAOYSA-N
SMILES: [Cl].N#CC1(CCNCC1)C1=CC=CC=C1
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)