accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 513-12-2

CH3SOOHO

CAS RN 513-12-2

IUPAC Name: 2-(ethanesulfonyl)ethan-1-ol
Synonyms: 2-(ethylsulfonyl)ethanol
Molecular Weight (g/mol): 138.18
Molecular Formula: C4H10O3S
InChi Key: LJNDRJPZYNGSKZ-UHFFFAOYSA-N
SMILES: CCS(=O)(=O)CCO
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results