accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 51264-70-1

H3COOOOOCH3

CAS RN 51264-70-1

IUPAC Name: ethyl 2-(4-formyl-2-methoxyphenoxy)acetate
Synonyms: ethyl 2-(4-formyl-2-methoxyphenoxy)acetate
Molecular Weight (g/mol): 238.24
Molecular Formula: C12H14O5
InChi Key: SVYWLZTVBMJCGG-UHFFFAOYSA-N
SMILES: CCOC(=O)COC1=C(OC)C=C(C=O)C=C1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results