accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 512181-73-6

NHOFF

CAS RN 512181-73-6

IUPAC Name: 4-fluoro-N-[(4-fluorophenyl)methyl]benzamide
Synonyms: 4-fluoro-N-[(4-fluorophenyl)methyl]benzamide
Molecular Weight (g/mol): 247.25
Molecular Formula: C14H11F2NO
InChi Key: CJFYBDORFMDXTG-UHFFFAOYSA-N
SMILES: FC1=CC=C(CNC(=O)C2=CC=C(F)C=C2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results