CAS RN 51116-01-9
CAS RN 51116-01-9
IUPAC Name:
9-[(4aR,6R,7R,7aS)-2,7-dihydroxy-2-oxo-hexahydro-2λ⁵-furo[3,2-d][1,3,2]dioxaphosphinin-6-yl]-2-amino-8-bromo-6,9-dihydro-3H-purin-6-one
Synonyms:
9-[(4aR,6R,7R,7aS)-2,7-dihydroxy-2-oxo-tetrahydro-4H-2λ⁵-furo[3,2-d][1,3,2]dioxaphosphinin-6-yl]-2-amino-8-bromo-3H-purin-6-one
Molecular Weight (g/mol):
424.1
Molecular Formula:
C10H11BrN5O7P
InChi Key:
YUFCOOWNNHGGOD-UMMCILCDSA-N
SMILES:
NC1=NC(=O)C2=C(N1)N([C@@H]1O[C@@H]3COP(O)(=O)O[C@H]3[C@H]1O)C(Br)=N2
1
–
1
of
1
results
1
Promotions Available
Tocris Bioscience™ 8-Bromo-cGMP, sodium salt
Tocris Bioscience™ 8-Bromo-cGMP, sodium salt
CAS: 51116-01-9 Molecular Formula: C10H11BrN5O7P