accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 51-80-9

IUPAC Name: [(dimethylamino)methyl]dimethylamine
Synonyms: [(dimethylamino)methyl]dimethylamine
Molecular Weight (g/mol): 102.18
Molecular Formula: C5H14N2
InChi Key: VGIVLIHKENZQHQ-UHFFFAOYSA-N
SMILES: CN(C)CN(C)C
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)