accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

H3CNNCH3OO

CAS RN 50920-65-5

IUPAC Name: 1-ethyl-3-methyl-1H-pyrazole-5-carboxylate
Synonyms: 2-ethyl-5-methylpyrazole-3-carboxylate
Molecular Weight (g/mol): 153.16
Molecular Formula: C7H9N2O2
InChi Key: VFMGOJUUTAPPDA-UHFFFAOYSA-M
SMILES: CCN1N=C(C)C=C1C([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results