accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 50877-42-4

INN

CAS RN 50877-42-4

IUPAC Name: 1-benzyl-4-iodo-1H-pyrazole
Synonyms: 1-benzyl-4-iodopyrazole
Molecular Weight (g/mol): 284.1
Molecular Formula: C10H9IN2
InChi Key: PVEYRBGIYMWFPB-UHFFFAOYSA-N
SMILES: IC1=CN(CC2=CC=CC=C2)N=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results