accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 50847-06-8

IUPAC Name: 2-phenyl-1H-imidazole-4,5-dicarbonitrile
Synonyms: 2-phenyl-1H-imidazole-4,5-dicarbonitrile
Molecular Weight (g/mol): 194.2
Molecular Formula: C11H6N4
InChi Key: PGSHQANDAMACEF-UHFFFAOYSA-N
SMILES: N#CC1=C(N=C(N1)C1=CC=CC=C1)C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results