accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CH3OOOCH3OCH3OH3CNOO

CAS RN 5081-42-5

IUPAC Name: methyl 3,4,5-trimethoxy-2-nitrobenzoate
Synonyms: methyl 3,4,5-trimethoxy-2-nitrobenzoate
Molecular Weight (g/mol): 271.23
Molecular Formula: C11H13NO7
InChi Key: HCEIEWYPDCDNNU-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC(OC)=C(OC)C(OC)=C1[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results