accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 50739-30-5

IUPAC Name: 2-benzyl-1,3-thiazolidine-4-carboxylic acid
Synonyms: 2-benzyl-1,3-thiazolidine-4-carboxylic acid
Molecular Weight (g/mol): 223.29
Molecular Formula: C11H13NO2S
InChi Key: SNFSDSLKMQQXDU-UHFFFAOYNA-N
SMILES: OC(=O)C1CSC(CC2=CC=CC=C2)N1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results