accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 5068-28-0

CH3CH3CH3OOHNSHOO

CAS RN 5068-28-0

IUPAC Name: 3-(benzylsulfanyl)-2-{[(tert-butoxy)carbonyl]amino}propanoic acid
Synonyms: 3-(benzylsulfanyl)-2-[(tert-butoxycarbonyl)amino]propanoic acid
Molecular Weight (g/mol): 311.4
Molecular Formula: C15H21NO4S
InChi Key: IFVORPLRHYROAA-UHFFFAOYNA-N
SMILES: CC(C)(C)OC(=O)NC(CSCC1=CC=CC=C1)C(O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)