accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 50594-78-0

IUPAC Name: 5-fluoro-2-nitrobenzonitrile
Synonyms: 5-fluoro-2-nitrobenzonitrile
Molecular Weight (g/mol): 166.11
Molecular Formula: C7H3FN2O2
InChi Key: VCEQYKYTIDJWTD-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=C(F)C=C1C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results