accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

OHNF

CAS RN 50543-23-2

IUPAC Name: 6-fluoro-1,2-dihydropyridin-2-one
Synonyms: 6-fluoro-1H-pyridin-2-one
Molecular Weight (g/mol): 113.09
Molecular Formula: C5H4FNO
InChi Key: AJRUXHIEYSYRQJ-UHFFFAOYSA-N
SMILES: FC1=CC=CC(=O)N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results