accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

FClNCO

CAS RN 50529-33-4

IUPAC Name: 2-chloro-1-fluoro-4-isocyanatobenzene
Synonyms: 2-chloro-1-fluoro-4-isocyanatobenzene
Molecular Weight (g/mol): 171.56
Molecular Formula: C7H3ClFNO
InChi Key: XVIPJBUXMFLHSI-UHFFFAOYSA-N
SMILES: FC1=CC=C(C=C1Cl)N=C=O
Boiling Point 
  • (3)
  • (1)
Molecular Weight (g/mol) 
  • (3)
  • (1)
Quantity 
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (3)
  • (1)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)