accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 5048-25-9

H2CN

CAS RN 5048-25-9

IUPAC Name: hept-6-enenitrile
Synonyms: hept-6-enenitrile
Molecular Weight (g/mol): 109.17
Molecular Formula: C7H11N
InChi Key: ZLWQKLRMIQIHKQ-UHFFFAOYSA-N
SMILES: C=CCCCCC#N
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)