accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 50458-79-2

H3COON

CAS RN 50458-79-2

IUPAC Name: ethyl isoquinoline-3-carboxylate
Synonyms: ethyl isoquinoline-3-carboxylate
Molecular Weight (g/mol): 201.23
Molecular Formula: C12H11NO2
InChi Key: IFSCYCNNAIADLI-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=NC=C2C=CC=CC2=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results