accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 50446-44-1

OOHOOHOOH

CAS RN 50446-44-1

IUPAC Name: 3',5'-bis(4-carboxyphenyl)-[1,1'-biphenyl]-4-carboxylic acid
Synonyms: 3',5'-bis(4-carboxyphenyl)-[1,1'-biphenyl]-4-carboxylic acid
Molecular Weight (g/mol): 438.44
Molecular Formula: C27H18O6
InChi Key: SATWKVZGMWCXOJ-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=C(C=C1)C1=CC(=CC(=C1)C1=CC=C(C=C1)C(O)=O)C1=CC=C(C=C1)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results