accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 504-31-4

OO

CAS RN 504-31-4

IUPAC Name: 2H-pyran-2-one
Synonyms: α-pyrone coumalin D-valerolactone
Molecular Weight (g/mol): 96.09
Molecular Formula: C5H4O2
InChi Key: ZPSJGADGUYYRKE-UHFFFAOYSA-N
SMILES: O=C1OC=CC=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results