accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 504-02-9

OO

CAS RN 504-02-9

IUPAC Name: cyclohexane-1,3-dione
Synonyms: 1,3 cyclohexanedione resorcinol, dihydro- 1,3-benzenediol, dihydro- dihydro-resorcinol 1,3-cyclohexanone dihydro-1,3-benzenediol
Molecular Weight (g/mol): 112.13
Molecular Formula: C6H8O2
InChi Key: HJSLFCCWAKVHIW-UHFFFAOYSA-N
SMILES: O=C1CCCC(=O)C1
Molecular Weight (g/mol) 
  • (4)
  • (4)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (4)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (4)
  • (1)
  • (4)
  • (4)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (4)
  • (3)
  • (3)