accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 5031-78-7

IUPAC Name: 1-(4-phenoxyphenyl)ethan-1-one
Synonyms: 1-(4-phenoxyphenyl)ethanone
Molecular Weight (g/mol): 212.25
Molecular Formula: C14H12O2
InChi Key: DJNIFZYQFLFGDT-UHFFFAOYSA-N
SMILES: CC(=O)C1=CC=C(OC2=CC=CC=C2)C=C1
Boiling Point 
  • (1)
  • (3)
Percent Purity 
  • (3)
  • (1)
Quantity 
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)