accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

ONNClClCl

CAS RN 50270-27-4

IUPAC Name: 2,4,6-trichloropyrimidine-5-carbaldehyde
Synonyms: 2,4,6-trichloropyrimidine-5-carbaldehyde
Molecular Weight (g/mol): 211.43
Molecular Formula: C5HCl3N2O
InChi Key: KVJIRFGNHAAUNQ-UHFFFAOYSA-N
SMILES: ClC1=NC(Cl)=C(C=O)C(Cl)=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results