accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 50269-95-9

IUPAC Name: 1H-pyrrole-2-carbohydrazide
Synonyms: 1H-pyrrole-2-carbohydrazide
Molecular Weight (g/mol): 125.13
Molecular Formula: C5H7N3O
InChi Key: FQIAWPZQJCAPSW-UHFFFAOYSA-N
SMILES: NNC(=O)C1=CC=CN1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results