accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

H3CONH2

CAS RN 50262-67-4

IUPAC Name: 4-(nonyloxy)aniline
Synonyms: 4-(nonyloxy)aniline
Molecular Weight (g/mol): 235.37
Molecular Formula: C15H25NO
InChi Key: JQLBBFVOAHUASD-UHFFFAOYSA-N
SMILES: CCCCCCCCCOC1=CC=C(N)C=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results