accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 5023-94-9

IUPAC Name: (Z)-N'-hydroxy-3-nitrobenzene-1-carboximidamide
Synonyms: (Z)-N'-hydroxy-3-nitrobenzenecarboximidamide
Molecular Weight (g/mol): 181.15
Molecular Formula: C7H7N3O3
InChi Key: ZAIHFKLUPWFUGH-UHFFFAOYSA-N
SMILES: N\C(=N/O)C1=CC=CC(=C1)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results