accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

HOO

CAS RN 5020-21-3

IUPAC Name: 1H-indene-1-carboxylic acid
Synonyms: 1H-indene-1-carboxylic acid
Molecular Weight (g/mol): 160.17
Molecular Formula: C10H8O2
InChi Key: KGWYICAEPBCRBL-UHFFFAOYNA-N
SMILES: OC(=O)C1C=CC2=CC=CC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results