accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

NHN

CAS RN 5004-07-9

IUPAC Name: 4-(pyrrolidin-1-yl)piperidine
Synonyms: 4-(pyrrolidin-1-yl)piperidine
Molecular Weight (g/mol): 154.26
Molecular Formula: C9H18N2
InChi Key: STWODXDTKGTVCJ-UHFFFAOYSA-N
SMILES: C1CCN(C1)C1CCNCC1
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results