accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 500346-91-8

CH3ClNHOCl

CAS RN 500346-91-8

IUPAC Name: 4-chloro-N-(3-chloro-2-methylphenyl)butanamide
Synonyms: 4-chloro-N-(3-chloro-2-methylphenyl)butanamide
Molecular Weight (g/mol): 246.13
Molecular Formula: C11H13Cl2NO
InChi Key: YTFOKDALOHUPSO-UHFFFAOYSA-N
SMILES: CC1=C(NC(=O)CCCCl)C=CC=C1Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results