accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

HOOOO

CAS RN 500-05-0

IUPAC Name: 2-oxo-2H-pyran-5-carboxylic acid
Synonyms: 6-oxopyran-3-carboxylic acid
Molecular Weight (g/mol): 140.09
Molecular Formula: C6H4O4
InChi Key: ORGPJDKNYMVLFL-UHFFFAOYSA-N
SMILES: OC(=O)C1=COC(=O)C=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results