accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

H3COOClH3C

CAS RN 499785-51-2

IUPAC Name: 2,2-dimethyl-2,3-dihydro-1-benzofuran-7-carbonyl chloride
Synonyms: 2,2-dimethyl-3H-1-benzofuran-7-carbonyl chloride
Molecular Weight (g/mol): 210.66
Molecular Formula: C11H11ClO2
InChi Key: LSSXEVHVRBTNAQ-UHFFFAOYSA-N
SMILES: CC1(C)CC2=CC=CC(C(Cl)=O)=C2O1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results