accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

H3CNONN

CAS RN 499770-67-1

IUPAC Name: 1-methyl-1H-1,2,3-benzotriazole-5-carbaldehyde
Synonyms: 1-methyl-1,2,3-benzotriazole-5-carbaldehyde
Molecular Weight (g/mol): 161.16
Molecular Formula: C8H7N3O
InChi Key: IPVPVOVAJDRRAK-UHFFFAOYSA-N
SMILES: CN1N=NC2=CC(C=O)=CC=C12
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results