accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 499104-69-7

IUPAC Name: 2-{[6-(benzyloxy)-7-acetamido-2-phenyl-hexahydro-2H-pyrano[3,2-d][1,3]dioxin-8-yl]oxy}propanoic acid
Synonyms: 2-{[6-(benzyloxy)-7-acetamido-2-phenyl-hexahydro-2H-pyrano[3,2-d][1,3]dioxin-8-yl]oxy}propanoic acid
Molecular Weight (g/mol): 471.51
Molecular Formula: C25H29NO8
InChi Key: JPPMVSNCFXDOJX-UHFFFAOYNA-N
SMILES: CC(OC1C(NC(C)=O)C(OCC2=CC=CC=C2)OC2COC(OC12)C1=CC=CC=C1)C(O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results