accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 49844-90-8

H3CSNClN

CAS RN 49844-90-8

IUPAC Name: 4-chloro-2-(methylsulfanyl)pyrimidine
Synonyms: 4-chloro-2-(methylsulfanyl)pyrimidine
Molecular Weight (g/mol): 160.62
Molecular Formula: C5H5ClN2S
InChi Key: DFOHHQRGDOQMKG-UHFFFAOYSA-N
SMILES: CSC1=NC=CC(Cl)=N1
Boiling Point 
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)