accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 498-60-2

OO

CAS RN 498-60-2

IUPAC Name: furan-3-carbaldehyde
Synonyms: furan-3-carboxaldehyde
Molecular Weight (g/mol): 96.09
Molecular Formula: C5H4O2
InChi Key: AZVSIHIBYRHSLB-UHFFFAOYSA-N
SMILES: O=CC1=COC=C1
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results