accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 49761-82-2

IUPAC Name: tert-butyl 1H-imidazole-1-carboxylate
Synonyms: tert-butyl imidazole-1-carboxylate
Molecular Weight (g/mol): 168.2
Molecular Formula: C8H12N2O2
InChi Key: MTBKGWHHOBJMHJ-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)N1C=CN=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results