accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

OHOO

CAS RN 4971-56-6

IUPAC Name: 5-hydroxy-2,5-dihydrofuran-2-one
Synonyms: tetronic acid
Molecular Weight (g/mol): 100.07
Molecular Formula: C4H4O3
InChi Key: DUAZKLYNTLDKQK-UHFFFAOYNA-N
SMILES: OC1OC(=O)C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results