accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

H3CCH3BrOHNNH2O

CAS RN 496-67-3

IUPAC Name: (2-bromo-3-methylbutanoyl)urea
Synonyms: 2-bromo-3-methylbutyrylurea α-bromo-I-valerylurea bromisoval (inn)
Molecular Weight (g/mol): 223.07
Molecular Formula: C6H11BrN2O2
InChi Key: CMCCHHWTTBEZNM-UHFFFAOYNA-N
SMILES: CC(C)C(Br)C(=O)NC(N)=O
Quantity 
  • (2)
  • (1)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results