accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 49572-60-3

H3CNNBr

CAS RN 49572-60-3

IUPAC Name: 2-bromo-1-methyl-1H-1,3-benzodiazole
Synonyms: 2-bromo-1-methyl-1,3-benzodiazole
Molecular Weight (g/mol): 211.06
Molecular Formula: C8H7BrN2
InChi Key: PROLKELCEPYEQT-UHFFFAOYSA-N
SMILES: CN1C(Br)=NC2=CC=CC=C12
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results