accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 495-18-1

IUPAC Name: N-hydroxybenzamide
Synonyms: benzhydroxamic acid benzohyroxamate
Molecular Weight (g/mol): 137.14
Molecular Formula: C7H7NO2
InChi Key: VDEUYMSGMPQMIK-UHFFFAOYSA-N
SMILES: ONC(=O)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results