accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 492-98-8

NHNHNN

CAS RN 492-98-8

IUPAC Name: 1H,1'H-2,2'-biimidazole
Synonyms: 2,2'-biimidazole
Molecular Weight (g/mol): 134.14
Molecular Formula: C6H6N4
InChi Key: AZUHIVLOSAPWDM-UHFFFAOYSA-N
SMILES: N1C=CN=C1C1=NC=CN1
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results