accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

SS

CAS RN 492-97-7

IUPAC Name: 2,2'-bithiophene
Synonyms: 2,2'-bithiophene α-bithiophene 2,2'-bithienyl dithienyl 2,2'-dithienyl
Molecular Weight (g/mol): 166.26
Molecular Formula: C8H6S2
InChi Key: OHZAHWOAMVVGEL-UHFFFAOYSA-N
SMILES: S1C=CC=C1C1=CC=CS1
Boiling Point 
  • (3)
  • (2)
Molecular Weight (g/mol) 
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)