accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CH3OOO

CAS RN 4919-33-9

IUPAC Name: 2-(4-ethoxyphenyl)acetate
Synonyms: 2-(4-ethoxyphenyl)acetate
Molecular Weight (g/mol): 179.2
Molecular Formula: C10H11O3
InChi Key: ZVVWZNFSMIFGEP-UHFFFAOYSA-M
SMILES: CCOC1=CC=C(CC([O-])=O)C=C1
Molecular Weight (g/mol) 
  • (2)
  • (3)
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (2)
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results